missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) adipate, 99%
Description
It is employed as a plasticizer. It is a skin conditioning solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a plasticizer. It is a skin conditioning solvent.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
It is employed as a plasticizer. It is a skin conditioning solvent.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -67°C |
| Density | 0.99 |
| Boiling Point | 166°C to 168°C (1 mmHg) |
| Flash Point | 196°C (384°F) |
| Refractive Index | 1.447 |
| Quantity | 25 g |
| Beilstein | 1803774 |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Formula Weight | 370.58 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H373 |
| Precautionary Statement | P260-P314-P501c |
| EINECSNumber | 203-090-1 |
| RTECSNumber | AU9700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only