missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) 3,3'-Thiodipropionate 96.0+%, TCI America™

Catalog No. p-7122938
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B342225G 25 g
B3422500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B342225G Supplier TCI America Supplier No. B342225G

Chemical Identifiers

CAS 10526-15-5
Molecular Formula C22H42O4S
Molecular Weight (g/mol) 402.634
MDL Number MFCD01757653
InChI Key NKNLSBYMGKTWDT-UHFFFAOYSA-N
PubChem CID 112063
IUPAC Name 2-ethylhexyl 3-[3-(2-ethylhexoxy)-3-oxopropyl]sulfanylpropanoate
SMILES CCCCC(CC)COC(=O)CCSCCC(=O)OCC(CC)CCCC

Specifications

Color Yellow
Boiling Point 180°C
Quantity 25 g
Synonym 3,3′C-Thiodipropionic Acid Bis(2-ethylhexyl) Ester
Formula Weight 402.63
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) 3,3′-Thiodipropionate
EINECSNumber (2)-1399
RTECSNumber UF7996250
TSCA Yes