CAS RN 10526-15-5
IUPAC Name:
2-ethylhexyl 3-({3-[(2-ethylhexyl)oxy]-3-oxopropyl}sulfanyl)propanoate
Synonyms:
2-ethylhexyl 3-({3-[(2-ethylhexyl)oxy]-3-oxopropyl}sulfanyl)propanoate
Molecular Weight (g/mol):
402.63
Molecular Formula:
C22H42O4S
InChi Key:
NKNLSBYMGKTWDT-UHFFFAOYNA-N
SMILES:
CCCCC(CC)COC(=O)CCSCCC(=O)OCC(CC)CCCC
1
–
1
of
1
results
1
Bis(2-ethylhexyl) 3,3'-Thiodipropionate 96.0+%, TCI America™
CAS: 10526-15-5 Molecular Formula: C22H42O4S Molecular Weight (g/mol): 402.634 MDL Number: MFCD01757653 InChI Key: NKNLSBYMGKTWDT-UHFFFAOYSA-N PubChem CID: 112063 IUPAC Name: 2-ethylhexyl 3-[3-(2-ethylhexoxy)-3-oxopropyl]sulfanylpropanoate SMILES: CCCCC(CC)COC(=O)CCSCCC(=O)OCC(CC)CCCC