missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexanoato)hydroxyaluminum 96.0+%, TCI America™
$29.83 - $149.13
Chemical Identifiers
| CAS | 30745-55-2 |
|---|---|
| Molecular Formula | C16H31AlO5 |
| Molecular Weight (g/mol) | 330.40 |
| MDL Number | MFCD00041940 |
| InChI Key | ITXFWNWNWXSDBB-UHFFFAOYNA-K |
| Synonym | Hydroxyaluminum Bis(2-ethylhexanoate) |
| IUPAC Name | aluminum(3+) bis(2-ethylhexanoate) hydroxide |
| SMILES | [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O |
Chemical Identifiers
| 30745-55-2 | |
| 330.40 | |
| ITXFWNWNWXSDBB-UHFFFAOYNA-K | |
| aluminum(3+) bis(2-ethylhexanoate) hydroxide |
| C16H31AlO5 | |
| MFCD00041940 | |
| Hydroxyaluminum Bis(2-ethylhexanoate) | |
| [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O |
Specifications
| 30745-55-2 | |
| C16H31AlO5 | |
| 25 g | |
| Hydroxyaluminum Bis(2-ethylhexanoate) | |
| [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O | |
| 330.40 | |
| ≥96.0% (T) | |
| Bis(2-ethylhexanoato)hydroxyaluminum |
| White | |
| MFCD00041940 | |
| 1325 | |
| ITXFWNWNWXSDBB-UHFFFAOYNA-K | |
| aluminum(3+) bis(2-ethylhexanoate) hydroxide | |
| 330.40 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes