missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexanoato)hydroxyaluminum 96.0+%, TCI America™
Supplier: TCI America B1895250G
Specifications
| Bis(2-ethylhexanoato)hydroxyaluminum | |
| White | |
| MFCD00041940 | |
| 1325 | |
| ITXFWNWNWXSDBB-UHFFFAOYNA-K | |
| aluminum(3+) bis(2-ethylhexanoate) hydroxide | |
| 330.40 | |
| Crystalline Powder |
| 30745-55-2 | |
| C16H31AlO5 | |
| 250 g | |
| Hydroxyaluminum Bis(2-ethylhexanoate) | |
| [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O | |
| 330.40 | |
| ≥96.0% (T) |
Chemical Identifiers
| 30745-55-2 | |
| 330.40 | |
| ITXFWNWNWXSDBB-UHFFFAOYNA-K | |
| aluminum(3+) bis(2-ethylhexanoate) hydroxide |
| C16H31AlO5 | |
| MFCD00041940 | |
| Hydroxyaluminum Bis(2-ethylhexanoate) | |
| [OH-].[Al+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O |
Safety and Handling
TSCA : Yes