missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%

Catalog No. AC316820050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC316820050 5 g Glass bottle
AC316820010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC316820050 Supplier Thermo Scientific Chemicals Supplier No. 316820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23582-02-7 | C34H33P3 | 534.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23582-02-7
Molecular Formula C34H33P3
Molecular Weight (g/mol) 534.56
MDL Number MFCD00003048
InChI Key AXVOAMVQOCBPQT-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinoethyl phenylphosphine, phenylphosphinediyl bis ethane-2,1-diyl bis diphenylphosphine, bis 2-diphenylphosphino ethyl phenylphosphine, phosphine, bis 2-diphenylphosphino ethyl phenyl, phenylbis diphenylphosphinoethyl phosphine, pubchem6540, acmc-20alo6, chembl69711, bis 2-biphenylphosphinoethyl phenylphosphine
PubChem CID 90192
IUPAC Name bis(2-diphenylphosphanylethyl)-phenylphosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 121.0°C to 126.0°C
Color White
Elemental Analysis 74.1 to 78.7% (C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 534.56
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material Bis(2-diphenylphosphinoethyl)phenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-753-8

RUO – Research Use Only