accessibility menu, dialog, popup

UserName

CAS RN 23582-02-7

IUPAC Name: (2-{[2-(diphenylphosphanyl)ethyl](phenyl)phosphanyl}ethyl)diphenylphosphane
Synonyms: triphos
Molecular Weight (g/mol): 534.56
Molecular Formula: C34H33P3
InChi Key: AXVOAMVQOCBPQT-UHFFFAOYSA-N
SMILES: C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)