missing translation for 'onlineSavingsMsg'
Learn More

Bis-(2-chloroisopropyl) ether, MilliporeSigma™ Supelco™

Catalog No. 111018161
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018161 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018161 Supplier Merck Emd Millipore Supplier No. 48498

Bis-(2-chloroisopropyl) ether is a colorless liquid, used as a solvent for greases, waxes and fats. It can also find applications in manufacturing of textiles, spotting agents and cleaning solutions.

Environmental

Specifications

Packaging Ampule
CAS 108-60-1
Flash Point 85°C
Physical Form Neat
Linear Formula C6H12 Cl2 O
MDL Number MFCD00272183
Synonym 2-Chloroisopropyl ether
Type Organic Pollutant Standard
Quantity 100 mg
Formula Weight 171.06
Grade Analytical Standard
Show More Show Less