missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) Carbonate 98.0+%, TCI America™

Catalog No. C003810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C003810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C003810G Supplier TCI America Supplier No. C003810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-97-2
Molecular Formula C5H8Cl2O3
Molecular Weight (g/mol) 187.016
MDL Number MFCD00018962
InChI Key WQULVXNWEYLDJY-UHFFFAOYSA-N
Synonym Carbonic Acid Bis(2-chloroethyl) Ester, Carbonic Acid Di(2-chloroethyl) Ester, Di(2-chloroethyl) Carbonate
PubChem CID 69345
IUPAC Name bis(2-chloroethyl) carbonate
SMILES C(CCl)OC(=O)OCCCl

Specifications

Color Yellow
Boiling Point 241°C
Quantity 10 g
Formula Weight 187.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-chloroethyl) Carbonate
RTECSNumber FF7900000
TSCA Yes