accessibility menu, dialog, popup

UserName

CAS RN 623-97-2

IUPAC Name: bis(2-chloroethyl) carbonate
Synonyms: bis(2-chloroethyl) carbonate
Molecular Weight (g/mol): 187.02
Molecular Formula: C5H8Cl2O3
InChi Key: WQULVXNWEYLDJY-UHFFFAOYSA-N
SMILES: ClCCOC(=O)OCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results