missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[2-carboxyethylgermanium(IV)] Sesquioxide 98.0+%, TCI America™
$160.30 - $527.52
Chemical Identifiers
| CAS | 12758-40-6 |
|---|---|
| Molecular Formula | C6H10Ge2O7 |
| Molecular Weight (g/mol) | 339.399 |
| MDL Number | MFCD00049319 |
| InChI Key | XEABSBMNTNXEJM-UHFFFAOYSA-N |
| Synonym | propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide |
| PubChem CID | 83030 |
| ChEBI | CHEBI:32060 |
| IUPAC Name | 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid |
| SMILES | C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O |
Chemical Identifiers
| 12758-40-6 | |
| 339.399 | |
| XEABSBMNTNXEJM-UHFFFAOYSA-N | |
| 83030 | |
| 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid |
| C6H10Ge2O7 | |
| MFCD00049319 | |
| propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide | |
| CHEBI:32060 | |
| C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O |
Specifications
| 12758-40-6 | |
| White-Yellow | |
| MFCD00049319 | |
| propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide | |
| C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O | |
| 339.399 | |
| CHEBI:32060 | |
| ≥98.0% (T) | |
| Bis[2-carboxyethylgermanium(IV)] Sesquioxide |
| 320°C | |
| C6H10Ge2O7 | |
| 5 g | |
| XEABSBMNTNXEJM-UHFFFAOYSA-N | |
| 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid | |
| 83030 | |
| 339.40 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UA2459750
TSCA : Yes