missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-aminophenyl) ether, 98%

Catalog No. p-7052699
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55396MD 250 mg
AAH5539603 1 g
AAH5539606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55396MD Supplier Thermo Scientific Chemicals Supplier No. H55396MD
Only null left

CAS: 24878-25-9 | C12H12N2O | 200.241 g/mol

It is catalytic reagent and used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is catalytic reagent and used in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Store at low temperature, away from light and moisture.

Chemical Identifiers

CAS 24878-25-9
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00792533
InChI Key GOJFAKBEASOYNM-UHFFFAOYSA-N
Synonym 2,2'-oxydianiline, 2-2-aminophenoxy aniline, 2,2 inverted exclamation marka-oxydianiline, aminophenyl ether, o,o'-oxydianiline, 2-aminophenyl ether, bis 2-aminophenyl ether, 2,2'-oxybis benzenamine, benzenamine,2,2'-oxybis, 2,2'-diaminodiphenyl ether
PubChem CID 458824
IUPAC Name 2-(2-aminophenoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC2=CC=CC=C2N

Specifications

Melting Point 61°C to 64°C
Color Brown to White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2-aminophenyl) ether
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only