missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,6-diisopropylphenyl)carbodiimide 98.0+%, TCI America™

Catalog No. B27565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27565G 5 g
B275625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27565G Supplier TCI America Supplier No. B27565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2162-74-5
Molecular Formula C25H34N2
Molecular Weight (g/mol) 362.561
InChI Key XLDBGFGREOMWSL-UHFFFAOYSA-N
PubChem CID 75100
IUPAC Name N,N'-bis[2,6-di(propan-2-yl)phenyl]methanediimine
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N=C=NC2=C(C=CC=C2C(C)C)C(C)C

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 362.56
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,6-diisopropylphenyl)carbodiimide
EINECSNumber (3)-0142
RTECSNumber CY0887250
TSCA Yes