missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)manganese(II) Dihydrate 97.0+%, TCI America™

$167.76 - $167.76

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M004225G
View Documents
TCI America
M004225G
25 g
Each for $167.76
Only null left
 
Description

Description

Specifications

Chemical Identifiers

14024-58-9
253.16
ZQZQURFYFJBOCE-FDGPNNRMSA-L
54669727
[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14MnO4
MFCD00000022 MFCD09998212
bis 4-hydroxypent-3-en-2-one dihydrate manganese
manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)

Specifications

14024-58-9
C10H14MnO4
25 g
ZQZQURFYFJBOCE-FDGPNNRMSA-L
manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
54669727
≥97.0% (T)
Bis(2,4-pentanedionato)manganese(II) Dihydrate
Yellow
MFCD00000022 MFCD09998212
bis 4-hydroxypent-3-en-2-one dihydrate manganese
[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
253.16
253.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2142

RTECSNumber : OO9350000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'