missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)magnesium(II) 98.0+%, TCI America™
$88.56 - $323.43
Chemical Identifiers
| CAS | 14024-56-7 |
|---|---|
| Molecular Formula | C10H14MgO4 |
| Molecular Weight (g/mol) | 222.523 |
| MDL Number | MFCD00013497 |
| InChI Key | AKTIAGQCYPCKFX-SYWGCQIGSA-L |
| Synonym | 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one |
| PubChem CID | 25113476 |
| IUPAC Name | magnesium;(E)-4-oxopent-2-en-2-olate |
| SMILES | CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2] |
Chemical Identifiers
| 14024-56-7 | |
| 222.523 | |
| AKTIAGQCYPCKFX-SYWGCQIGSA-L | |
| 25113476 | |
| CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2] |
| C10H14MgO4 | |
| MFCD00013497 | |
| 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one | |
| magnesium;(E)-4-oxopent-2-en-2-olate |
Specifications
| 14024-56-7 | |
| White | |
| MFCD00013497 | |
| 4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one | |
| CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2] | |
| 222.523 | |
| 222.52 | |
| Crystalline Powder |
| 276°C | |
| C10H14MgO4 | |
| 5 g | |
| AKTIAGQCYPCKFX-SYWGCQIGSA-L | |
| magnesium;(E)-4-oxopent-2-en-2-olate | |
| 25113476 | |
| ≥98.0% (T) | |
| Bis(2,4-pentanedionato)magnesium(II) |
Safety and Handling
EINECSNumber : (2)-2181
TSCA : No