accessibility menu, dialog, popup

UserName

CAS RN 14024-56-7

IUPAC Name: magnesium(2+) bis(2,4-dioxopentan-3-ide)
Synonyms: magnesium(2+) diacetylacetonate
Molecular Weight (g/mol): 222.52
Molecular Formula: C10H14MgO4
InChi Key: QTNPNISUJKZVIS-UHFFFAOYSA-N
SMILES: [Mg++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results