missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,4-pentanedionato)copper(II) 97.0+%, TCI America™
Supplier: TCI America C038425G
Specifications
| Bis(2,4-pentanedionato)copper(II) | |
| 288°C | |
| C10H14CuO4 | |
| 25 g | |
| QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
| copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
| 261.76 |
| 13395-16-9 | |
| 160°C | |
| MFCD00000016 | |
| Acetylacetone Copper(II) Salt, Copper(II) Acetylacetonate, Cupric Acetylacetonate | |
| [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
| 261.76 | |
| ≥97.0% (T) |
Chemical Identifiers
| 13395-16-9 | |
| 261.76 | |
| QYJPSWYYEKYVEJ-FDGPNNRMSA-L | |
| copper(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
| C10H14CuO4 | |
| MFCD00000016 | |
| Acetylacetone Copper(II) Salt, Copper(II) Acetylacetonate, Cupric Acetylacetonate | |
| [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Safety and Handling
EINECSNumber : (2)-2142&(2)-2181
RTECSNumber : GL6520000
TSCA : Yes