missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)copper(II) 97.0+%, TCI America™

Catalog No. p-7124387
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C038425G 25 g
C0384250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C038425G Supplier TCI America Supplier No. C038425G
Only null left

Chemical Identifiers

CAS 13395-16-9
Molecular Formula C10H14CuO4
Molecular Weight (g/mol) 261.76
MDL Number MFCD00000016
InChI Key QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Synonym Acetylacetone Copper(II) Salt, Copper(II) Acetylacetonate, Cupric Acetylacetonate
IUPAC Name copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 288°C
Boiling Point 160°C
Quantity 25 g
Formula Weight 261.76
Percent Purity ≥97.0% (T)
Chemical Name or Material Bis(2,4-pentanedionato)copper(II)
EINECSNumber (2)-2142&(2)-2181
RTECSNumber GL6520000
TSCA Yes