missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-pentanedionato)calcium(II) 98.0+%, TCI America™

Catalog No. C093025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C093025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C093025G Supplier TCI America Supplier No. C093025G
Only null left

Chemical Identifiers

CAS 19372-44-2
Molecular Formula C10H18CaO6
Molecular Weight (g/mol) 274.326
MDL Number MFCD00013486
InChI Key FTDUGYDPOWCKTD-VGKOASNMSA-L
Synonym Acetylacetone Calcium(II) Salt, Calcium(II) Acetylacetonate
PubChem CID 131675864
IUPAC Name calcium;(Z)-4-oxopent-2-en-2-olate;dihydrate
SMILES CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.O.[Ca+2]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 238.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4-pentanedionato)calcium(II)
EINECSNumber (2)-2181
TSCA Yes