accessibility menu, dialog, popup

UserName

CaCH3OCHCH3OCH3OCHCH3O

CAS RN 19372-44-2

IUPAC Name: calcium bis(2,4-dioxopentan-3-ide)
Synonyms: calcium diacetylacetonate
Molecular Weight (g/mol): 238.3
Molecular Formula: C10H14CaO4
InChi Key: XVYOQBDTLDXFQL-UHFFFAOYSA-N
SMILES: [Ca++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results