missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4,6-trimethylpyridine)iodonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. B23591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23591G 1 g
B23595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23591G Supplier TCI America Supplier No. B23591G

Chemical Identifiers

CAS 113119-46-3
Molecular Formula C16H22IN2
Molecular Weight (g/mol) 369.27
MDL Number MFCD00799490
InChI Key VULYHYQCXMNHOI-UHFFFAOYSA-N
IUPAC Name bis(2,4,6-trimethylpyridin-1-ium-1-yl)iodanuide
SMILES CC1=CC(C)=[N+]([I-][N+]2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 1 g
Formula Weight 514.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4,6-trimethylpyridine)iodonium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator