accessibility menu, dialog, popup

UserName

CH3CH3NINCH3CH3H3CH3C

CAS RN 113119-46-3

IUPAC Name: bis(2,4,6-trimethylpyridin-1-ium-1-yl)iodanuide
Synonyms: bis(2,4,6-trimethylpyridin-1-ium-1-yl)iodanuide
Molecular Weight (g/mol): 369.27
Molecular Formula: C16H22IN2
InChi Key: VULYHYQCXMNHOI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=[N+]([I-][N+]2=C(C)C=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results