missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4,6-trichlorophenyl) Oxalate 98.0+%, TCI America™

Catalog No. p-7118269
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53021G 1 g
A53025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53021G Supplier TCI America Supplier No. A53021G
Only null left

[Chemiluminescence reagent for the determination of fluorescent compounds]

Chemical Identifiers

CAS 1165-91-9
Molecular Formula C14H4Cl6O4
Molecular Weight (g/mol) 448.882
MDL Number MFCD00043061
InChI Key GEVPIWPYWJZSPR-UHFFFAOYSA-N
Synonym bis 2,4,6-trichlorophenyl oxalate, bis 2,4,6-trichlorophenyl ethanedioate, oxalic acid bis 2,4,6-trichlorophenyl ester, 2,4,6-trichlorophenyl oxalate, ethanedioic acid, bis 2,4,6-trichlorophenyl ester, bis-tcpo, acmc-20a7jt, gevpiwpywjzspr-uhfffaoysa
PubChem CID 160567
IUPAC Name bis(2,4,6-trichlorophenyl) oxalate
SMILES C1=C(C=C(C(=C1Cl)OC(=O)C(=O)OC2=C(C=C(C=C2Cl)Cl)Cl)Cl)Cl

Specifications

Melting Point 190°C
Color White
Quantity 1 g
Formula Weight 448.88
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,4,6-trichlorophenyl) Oxalate [Chemiluminescence reagent for the determination of fluorescent c