missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,2-diethoxyethyl) Disulfide 90.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 155°C |
| Quantity | 1 g |
| Formula Weight | 298.46 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(2,2-diethoxyethyl) Disulfide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |