missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(2-benzoxazolyl)phenolato]zinc(II) 97.0+%, TCI America™

Supplier:  TCI America B20785G

Catalog No. B20785G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis[2-(2-benzoxazolyl)phenolato]zinc(II)
C26H18N2O4Zn
5 g
C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2.[Zn]
487.82
485.80
Crystalline Powder
23467-27-8
MFCD03844767
KYYRCGSPILJUQZ-UHFFFAOYSA-N
6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc
132773703
≥97.0% (T)

Chemical Identifiers

23467-27-8
487.82
KYYRCGSPILJUQZ-UHFFFAOYSA-N
6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc
C26H18N2O4Zn
MFCD03844767
132773703
C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)O2.[Zn]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions