accessibility menu, dialog, popup

UserName

CAS RN 23467-27-8

IUPAC Name: bis(6-(2,3-dihydro-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one) zinc
Synonyms: bis(6-(3H-1,3-benzoxazol-2-ylidene)cyclohexa-2,4-dien-1-one) zinc
Molecular Weight (g/mol): 487.82
Molecular Formula: C26H18N2O4Zn
InChi Key: KYYRCGSPILJUQZ-UHFFFAOYSA-N
SMILES: [Zn].O=C1C=CC=CC1=C1NC2=CC=CC=C2O1.O=C1C=CC=CC1=C1NC2=CC=CC=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results