missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(2-benzothiazolyl)phenolato]zinc(II) 98.0+%, TCI America™

Catalog No. B20775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20775G 5 g
B20771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20775G Supplier TCI America Supplier No. B20775G
Only null left

Chemical Identifiers

CAS 58280-31-2
Molecular Formula C26H18N2O2S2Zn
Molecular Weight (g/mol) 519.942
MDL Number MFCD03844766
InChI Key MPDFTJIWQKPYAF-YJCYQSLBSA-N
PubChem CID 50878549
IUPAC Name (6E)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc
SMILES C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.[Zn]

Specifications

Quantity 5 g
Formula Weight 517.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[2-(2-benzothiazolyl)phenolato]zinc(II)