missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate 98.0+%, TCI America™

Supplier:  TCI America S044825G

 View more versions of this product

Catalog No. S044825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate
83°C
C28H52N2O4
25 g
bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
480.73
480.73
Crystalline Powder
52829-07-9
White
MFCD00134709
2811
RWMYPXKVMUFMKS-UHFFFAOYSA-N
bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
164282
≥98.0% (GC)

Chemical Identifiers

52829-07-9
480.73
RWMYPXKVMUFMKS-UHFFFAOYSA-N
164282
CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
C28H52N2O4
MFCD00134709
bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3732

RTECSNumber : HD8315000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions