missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate 98.0+%, TCI America™

Catalog No. p-7135891
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Catalog No. Quantity
S04485G 5 g
S044825G 25 g
S0448250G 250 g
3 options

Catalog No. S04485G

Supplier: TCI America S04485G

Only null left

Chemical Identifiers

CAS 52829-07-9
Molecular Formula C28H52N2O4
Molecular Weight (g/mol) 480.73
MDL Number MFCD00134709
InChI Key RWMYPXKVMUFMKS-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-4-piperidyl sebacate, bis 2,2,6,6-tetramethylpiperidin-4-yl decanedioate, tinuvin 770, sanol, sumisorb 577, tinuvin 770df, tinuvin 770ls, sanol 770, sanol ls 700, sanol ls 770
PubChem CID 164282
IUPAC Name bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
SMILES CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C

Specifications

Melting Point 83°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 480.73
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-4-piperidyl) Sebacate
EINECSNumber (5)-3732
RTECSNumber HD8315000
TSCA Yes