missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%

Catalog No. AA4047003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4047003 1 g
AA4047006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4047003 Supplier Thermo Scientific Chemicals Supplier No. 04047003

NMR shift reagent, nickel oxide precursor | CAS: 14481-08-4 | C22H38NiO4 | 425.24 g/mol

NMR shift reagent, nickel oxide precursor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14481-08-4
Molecular Formula C22H38NiO4
Molecular Weight (g/mol) 425.24
MDL Number MFCD00192348
InChI Key FPQQRRXEOLXSCJ-UHFFFAOYSA-N
Synonym bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2
PubChem CID 131675868
IUPAC Name (E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel
SMILES [Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 223°C to 225°C
Boiling Point 90°C to 110°C (0.1 mmHg)(sublimation)
Quantity 1 g
Formula Weight 425.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)
Hazard Category H317-H350
Hazard Statement GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only