missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate 95.0+%, TCI America™
$98.18 - $1305.26
Chemical Identifiers
| CAS | 88738-78-7 |
|---|---|
| Molecular Formula | C7H9F6O5P |
| Molecular Weight (g/mol) | 318.108 |
| MDL Number | MFCD00009901 |
| InChI Key | PVSJXEDBEXYLML-UHFFFAOYSA-N |
| Synonym | methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4 |
| PubChem CID | 184894 |
| IUPAC Name | methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate |
| SMILES | COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F |
Chemical Identifiers
| 88738-78-7 | |
| 318.108 | |
| PVSJXEDBEXYLML-UHFFFAOYSA-N | |
| 184894 | |
| COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F |
| C7H9F6O5P | |
| MFCD00009901 | |
| methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4 | |
| methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate |
Specifications
| 88738-78-7 | |
| C7H9F6O5P | |
| 1 g | |
| PVSJXEDBEXYLML-UHFFFAOYSA-N | |
| methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate | |
| 184894 | |
| ≥95.0% (GC) | |
| Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate |
| Yellow | |
| MFCD00009901 | |
| methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4 | |
| COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F | |
| 318.108 | |
| 318.11 | |
| Liquid |
Safety and Handling
TSCA : No