missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate 95.0+%, TCI America™

Supplier:  TCI America B17145G

 View more versions of this product

Catalog No. B17145G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate
Yellow
MFCD00009901
methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4
COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
318.108
318.11
Liquid
88738-78-7
C7H9F6O5P
5 g
PVSJXEDBEXYLML-UHFFFAOYSA-N
methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
184894
≥95.0% (GC)

Chemical Identifiers

88738-78-7
318.108
PVSJXEDBEXYLML-UHFFFAOYSA-N
184894
COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
C7H9F6O5P
MFCD00009901
methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4
methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions