missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,2,2-trifluoroethyl) Carbonate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 118°C |
| Quantity | 5 g |
| UN Number | 3272 |
| Formula Weight | 226.07 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(2,2,2-trifluoroethyl) Carbonate |
Safety and Handling
| TSCA | No |