CAS RN 1513-87-7
IUPAC Name:
bis(2,2,2-trifluoroethyl) carbonate
Synonyms:
bis(2,2,2-trifluoroethyl) carbonate
Molecular Weight (g/mol):
226.07
Molecular Formula:
C5H4F6O3
InChi Key:
WLLOZRDOFANZMZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)COC(=O)OCC(F)(F)F
1
–
1
of
1
results
1
Bis(2,2,2-trifluoroethyl) Carbonate 98.0+%, TCI America™
CAS: 1513-87-7 Molecular Formula: C5H4F6O3 Molecular Weight (g/mol): 226.074 MDL Number: MFCD11857845 InChI Key: WLLOZRDOFANZMZ-UHFFFAOYSA-N Synonym: Carbonic Acid Bis(2,2,2-trifluoroethyl) Ester PubChem CID: 11447463 IUPAC Name: bis(2,2,2-trifluoroethyl) carbonate SMILES: C(C(F)(F)F)OC(=O)OCC(F)(F)F