Learn More
Bis(2,2,2-trifluoroethyl)amine, 95%
CAS: 407-01-2 | C4H5F6N | 181.081 g/mol
Supplier: Thermo Scientific Chemicals L1670906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bis(2,2,2-trifluoroethyl)amine | |
| 1.1 | |
| C4H5F6N | |
| MFCD00042094 | |
| UN2735 | |
| bis 2,2,2-trifluoroethyl amine, bis trifluoroethyl amine, 1,1,1,5,5,5-hexafluoro-3-azapentane, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl ethanamine, ethanamine, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl, bis trifluoroethyl-amin, acmc-1aha1, 1,1,15,5,5-hexafluoro-3-azapentane, ethanamine,2,2,2-trifluoro-n-2,2,2-trifluoroethyl, 2,2,2-tris fluoranyl-n-2,2,2-tris fluoranyl ethyl ethanamine | |
| C(C(F)(F)F)NCC(F)(F)F | |
| 181.081 | |
| 181.08 |
| 407-01-2 | |
| 70°C to 71°C | |
| 1.301 | |
| 5 g | |
| 1811926 | |
| GTJGHXLFPMOKCE-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine | |
| 78999 | |
| 96% |
Chemical Identifiers
| 407-01-2 | |
| 181.081 | |
| GTJGHXLFPMOKCE-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine |
| C4H5F6N | |
| MFCD00042094 | |
| 78999 | |
| C(C(F)(F)F)NCC(F)(F)F |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 206-978-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only