missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl)amine, 95%

Catalog No. p-7046892
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1670903 1 g
AAL1670906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1670903 Supplier Thermo Scientific Chemicals Supplier No. L1670903

CAS: 407-01-2 | C4H5F6N | 181.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 407-01-2
Molecular Formula C4H5F6N
Molecular Weight (g/mol) 181.081
MDL Number MFCD00042094
InChI Key GTJGHXLFPMOKCE-UHFFFAOYSA-N
PubChem CID 78999
IUPAC Name 2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine
SMILES C(C(F)(F)F)NCC(F)(F)F

Specifications

Density 1.1
Boiling Point 70°C to 71°C
Refractive Index 1.301
Quantity 1 g
UN Number UN2735
Beilstein 1811926
Synonym bis 2,2,2-trifluoroethyl amine, bis trifluoroethyl amine, 1,1,1,5,5,5-hexafluoro-3-azapentane, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl ethanamine, ethanamine, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl, bis trifluoroethyl-amin, acmc-1aha1, 1,1,15,5,5-hexafluoro-3-azapentane, ethanamine,2,2,2-trifluoro-n-2,2,2-trifluoroethyl, 2,2,2-tris fluoranyl-n-2,2,2-tris fluoranyl ethyl ethanamine
Formula Weight 181.08
Percent Purity 96%
Chemical Name or Material Bis(2,2,2-trifluoroethyl)amine
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 206-978-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only