missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) Tetrafluoroborate, TCI America™

Catalog No. B39611G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39611G 1 g
B3961100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39611G Supplier TCI America Supplier No. B39611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35138-22-8
Molecular Formula C16H24BF4Rh
Molecular Weight (g/mol) 406.08
MDL Number MFCD00075045
InChI Key NBGSCPNNKGVTKU-QMDOQEJBSA-N
Synonym bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
PubChem CID 5702662
IUPAC Name λ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) tetrafluoroboranuide
SMILES [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 184°C
Quantity 1 g
UN Number 1759
Formula Weight 406.08
Physical Form Crystalline Powder
Chemical Name or Material Bis(1,5-cyclooctadiene)rhodium(I) Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator