accessibility menu, dialog, popup

UserName

CAS RN 35138-22-8

IUPAC Name: λ¹-rhodium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) tetrafluoroboranuide
Synonyms: λ¹-rhodium(1+) bis(1,5-cyclooctadiene, (Z,Z)-) tetrafluoroborate
Molecular Weight (g/mol): 406.08
Molecular Formula: C16H24BF4Rh
InChi Key: NBGSCPNNKGVTKU-QMDOQEJBSA-N
SMILES: [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)