missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Sebacate 95.0+%, TCI America™

Catalog No. p-7123248
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39245G 5 g
B392425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39245G Supplier TCI America Supplier No. B39245G

Chemical Identifiers

CAS 41556-26-7
Molecular Formula C30H56N2O4
Molecular Weight (g/mol) 508.788
MDL Number MFCD00134706
InChI Key RSOILICUEWXSLA-UHFFFAOYSA-N
Synonym Sebacic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester, Decanedioic Acid Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Ester
PubChem CID 586744
IUPAC Name bis(1,2,2,6,6-pentamethylpiperidin-4-yl) decanedioate
SMILES CC1(CC(CC(N1C)(C)C)OC(=O)CCCCCCCCC(=O)OC2CC(N(C(C2)(C)C)C)(C)C)C

Specifications

Color Yellow
Boiling Point 222°C
Quantity 5 g
Formula Weight 508.79
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(1,2,2,6,6-pentamethyl-4-piperidyl) Sebacate
EINECSNumber (5)-5501
TSCA Yes