accessibility menu, dialog, popup

UserName

CAS RN 41556-26-7

IUPAC Name: bis(3,3,4,5,5-pentamethylpiperidin-4-yl) decanedioate
Synonyms: pentamethyl piperidinyl sesquisebacate
Molecular Weight (g/mol): 508.79
Molecular Formula: C30H56N2O4
InChi Key: WHYLJCKAUWZIIS-UHFFFAOYSA-N
SMILES: CC1(C)CNCC(C)(C)C1(C)OC(=O)CCCCCCCCC(=O)OC1(C)C(C)(C)CNCC1(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results