missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl Zone Refined (number of passes:24) 99.0+%, TCI America™
Supplier: TCI America B02241SAMPLE
Specifications
| Biphenyl Zone Refined (number of passes:24) | |
| 70°C | |
| 256°C | |
| MFCD00003054 | |
| 3077 | |
| ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
| 1,1'-biphenyl | |
| 7095 | |
| 154.21 | |
| Crystalline Powder |
| 92-52-4 | |
| White | |
| C12H10 | |
| ZONE | |
| biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
| C1=CC=C(C=C1)C1=CC=CC=C1 | |
| 154.21 | |
| CHEBI:17097 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 92-52-4 | |
| 154.21 | |
| ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
| 7095 | |
| 1,1'-biphenyl |
| C12H10 | |
| MFCD00003054 | |
| biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
| CHEBI:17097 | |
| C1=CC=C(C=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (4)-0013
RTECSNumber : DU8050000
TSCA : Yes