missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl 99.5+%, TCI America™

Catalog No. B046525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B046525G 25 g
B0465500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B046525G Supplier TCI America Supplier No. B046525G

Chemical Identifiers

CAS 92-52-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003054
InChI Key ZUOUZKKEUPVFJK-UHFFFAOYSA-N
Synonym biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al
PubChem CID 7095
ChEBI CHEBI:17097
IUPAC Name 1,1'-biphenyl
SMILES C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 256°C
Quantity 25 g
UN Number 3077
Formula Weight 154.21
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl
EINECSNumber (4)-0013
RTECSNumber DU8050000
TSCA Yes