missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 68.0°C to 72.0°C |
| Color | White to Yellow |
| Boiling Point | 255.0°C |
| Flash Point | 113°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Plastic drum |
| Linear Formula | C6H5C6H5 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/att |
| EINECSNumber | 202-163-5 |
RUO – Research Use Only