missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl, 99%

Catalog No. p-7080649
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA102650B 1000 g
AAA102650I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA102650B Supplier Thermo Scientific Chemicals Supplier No. A102650B
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 92-52-4 | C12H10 | 154.21 g/mol

Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

Solubility
Soluble in alcohol and ether

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.

Chemical Identifiers

CAS 92-52-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003054
InChI Key ZUOUZKKEUPVFJK-UHFFFAOYSA-N
Synonym biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al
PubChem CID 7095
ChEBI CHEBI:17097
IUPAC Name 1,1'-biphenyl
SMILES C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 69°C to 72°C
Density 0.99
Boiling Point 254°C to 255°C
Flash Point 113°C (235°F)
Odor Pleasant
Quantity 1000 g
UN Number UN3077
Beilstein 1634058
Merck Index 14,3314
Solubility Information Soluble in alcohol and ether
Formula Weight 154.21
Percent Purity 99%
Chemical Name or Material Biphenyl
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-163-5
RTECSNumber DU8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only