missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134875
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P096125G 25 g
P0961100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P096125G Supplier TCI America Supplier No. P096125G

Chemical Identifiers

CAS 92-92-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002553
InChI Key NNJMFJSKMRYHSR-UHFFFAOYSA-N
Synonym 4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
PubChem CID 66724
IUPAC Name 4-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 227°C
Color White
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-4-carboxylic Acid
EINECSNumber (4)-0022
RTECSNumber DV1925100
TSCA Yes