missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7134838
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P090625G 25 g
P0906100G 100 g
P0906500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P090625G Supplier TCI America Supplier No. P090625G

Chemical Identifiers

CAS 947-84-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002463
InChI Key ILYSAKHOYBPSPC-UHFFFAOYSA-N
Synonym 2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
PubChem CID 70357
IUPAC Name 2-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

Specifications

Melting Point 113°C
Boiling Point 344°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-2-carboxylic Acid