missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-carboxylic acid, 98%

CAS: 947-84-2 | C13H10O2 | 198.221 g/mol

Supplier:  Thermo Scientific Chemicals A1204922

 View more versions of this product

Catalog No. AAA1204922

Only null left
Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate. he reaction of 2?-substituted biphenyl-2-carboxylic acids (where the 2?-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. he reaction of 2′-substituted biphenyl-2-carboxylic acids (where the 2′-substituent is H, CO2H, NO2, Cl, OMe, or CO2Me) with lead tetra-acetate in refluxing benzene solution, under a nitrogen atmosphere, affords 3,4-benzocoumarin as a major organic product.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

Biphenyl-2-carboxylic acid
110°C to 114°C
Odorless
MFCD00002463
974075
Insoluble in water.
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O
198.221
198.22
947-84-2
343°C to 344°C
C13H10O2
100 g
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
ILYSAKHOYBPSPC-UHFFFAOYSA-N
2-phenylbenzoic acid
70357
98%

Chemical Identifiers

947-84-2
198.221
ILYSAKHOYBPSPC-UHFFFAOYSA-N
70357
C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O
C13H10O2
MFCD00002463
2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
2-phenylbenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-432-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only