Learn More
Sartorius Biotrate Digital Burets
For trouble-free and reliable liquid dispensing directly from a supply bottle. Sartorius Biotrate Digital Burets offer high chemical resistance and is a smart solution for titrating various liquids accurately and safely.
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Color | White |
| Material | PTFE |
| Includes | Bottle Adapters: A32, A38, S40 |
Description
Biotrate is a premium digital burette equipped with sophisticated functions for easy titration. Biotrate is easy and convenient to use due to its large and clear electronic display, smooth operating wheels, and 360° rotating dispensing head.
- Smooth and light to operate with soft touch operating wheels
- Media recirculation system that allows unused media to be recovered from device and returned to bottle
- 360° Rotating dispensing head
- Universal connector at back of unit for drying tubes and filters
- Easy to attach aspiration tube with secure threaded connection User replaceable valves
- Supplied with UV-protected replacement window for light sensitive media protection
- Long life CR2032 battery
- Easy custom adjustment for various liquids
- Autoclavable liquid pathway
- Wide range of adapters included to fit most common bottle sizes
Specifications
| Autoclavable | |
| White | |
| PTFE | |
| Bottle Adapters: A32, A38, S40 |