missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin Recombinant Mouse Monoclonal Antibody (mAb2)
GREENER_CHOICE

Catalog No. MA551924
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551924 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551924 Supplier Invitrogen™ Supplier No. MA551924
Only null left

Mouse Recombinant Monoclonal Antibody

This antibody is capable of recognizing biotin conjugated proteins. It has very little or no affinity towards binding free biotin.

Biotin is a water-soluble B complex vitamin, generally classified as Vitamin B7. It is required by all organisms and can only be synthesized by bacteria, yeasts, molds, algae, and some plant species. Biotin is required for incorporation of carbon dioxide into organic compounds and exhibits a MW of ~244 Da.

Specifications

Antigen Biotin
Applications ELISA
Classification Recombinant Monoclonal
Clone mAb2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Vitamin B4
Host Species Mouse
Immunogen The original antibody was generated by immunizing femal BALB/c mice with biotin-KLH conjugate.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less