missing translation for 'onlineSavingsMsg'
Learn More

Biotin-PEG6-Maleimide 96.0+%, TCI America™

Catalog No. p-7124118
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B556310MG 10 mg
B556350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B556310MG Supplier TCI America Supplier No. B556310MG
Only null left

Chemical Identifiers

CAS 1808990-66-0
Molecular Formula C31H51N5O11S
Molecular Weight (g/mol) 701.833
InChI Key ILVNKONXZBFCPD-QRQMUESOSA-N
Synonym N-Biotinyl-N′-(3-maleimidopropionyl)-3,6,9,12,15,18-hexaoxaicosane-1,20-diamine, N-[2-[2-[2-[2-[2-[2-[2-[(3-Maleimidopropionyl)amino]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]biotinamide
PubChem CID 77078215
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-[2-[2-[2-[2-[3-(2,5-dioxopyrrol-1-yl)propanoylamino]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCOCCOCCOCCNC(=O)CCN3C(=O)C=CC3=O)NC(=O)N2

Specifications

Color White
Quantity 10 mg
Formula Weight 701.83
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin-PEG6-Maleimide
TSCA No
Recommended Storage Freezer (-20 °C)